Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetramethylammonium tetrafluoroborate | 661-36-9 | MFCD00011745 | ≥98.0% | 160.95 g/mol | C4H12BF4N | 10 G
SDP

Catalog No. 5000282776
Encompass_Preferred

Tetramethylammonium tetrafluoroborate is a high-purity biochemical reagent used as a source of the tetramethylammonium cation in organic synthesis, electrochemical studies, and analytical methods. The material is supplied as a crystalline salt with supporting documentation for research and laboratory use.

  • High purity ≥98.0% for reproducible experimental results.
  • Molecular weight 160.95 g/mol for accurate stoichiometry calculations.
  • Chemical formula C4H12BF4N for identity confirmation.
  • Available in convenient lab quantities, including 10 G packaging.
  • Stored sealed and protected from moisture; cold storage recommended for solutions.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.

Catalog No. 50-002-82776 Supplier Medchemexpress LLC Supplier No. HYW12767210G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetramethylammonium tetrafluoroborate is a high-purity biochemical reagent used as a source of the tetramethylammonium cation in organic synthesis, electrochemical studies, and analytical methods. The material is supplied as a crystalline salt with supporting documentation for research and laboratory use.

  • High purity ≥98.0% for reproducible experimental results.
  • Molecular weight 160.95 g/mol for accurate stoichiometry calculations.
  • Chemical formula C4H12BF4N for identity confirmation.
  • Available in convenient lab quantities, including 10 G packaging.
  • Stored sealed and protected from moisture; cold storage recommended for solutions.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.