Learn More
Medchemexpress LLC Tetramethylammonium tetrafluoroborate | 661-36-9 | MFCD00011745 | 98.0% | 160.95 g/mol | C4H12BF4N | 100 G
SDP
Catalog No. 5000283625
Encompass_Preferred
Tetramethylammonium tetrafluoroborate is a reagent used in chemical and biochemical research as a source of tetramethylammonium cations paired with tetrafluoroborate anions. It is supplied as a solid suitable for synthesis, electrochemistry, and analytical applications.
  • Provides tetramethylammonium cation with tetrafluoroborate anion for synthetic and electrochemical use.
  • High purity: 98.0%.
  • Molecular weight 160.95 g/mol; formula C4H12BF4N.
  • Available in multiple sizes including 100 G for laboratory use.
  • Stable when stored sealed at 4°C and protected from moisture.
  • Manufacturer documentation available for handling and quality (datasheet, COA, SDS).
Catalog No. 50-002-83625 Supplier Medchemexpress LLC Supplier No. HYW127672100G
Tetramethylammonium tetrafluoroborate is a reagent used in chemical and biochemical research as a source of tetramethylammonium cations paired with tetrafluoroborate anions. It is supplied as a solid suitable for synthesis, electrochemistry, and analytical applications.
  • Provides tetramethylammonium cation with tetrafluoroborate anion for synthetic and electrochemical use.
  • High purity: 98.0%.
  • Molecular weight 160.95 g/mol; formula C4H12BF4N.
  • Available in multiple sizes including 100 G for laboratory use.
  • Stable when stored sealed at 4°C and protected from moisture.
  • Manufacturer documentation available for handling and quality (datasheet, COA, SDS).