Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetramethylammonium tetrafluoroborate | 661-36-9 | MFCD00011745 | 98.0% | 160.95 g/mol | C4H12BF4N | 50 G
SDP

Catalog No. 5000282745
Encompass_Preferred

Tetramethylammonium tetrafluoroborate is an inorganic quaternary ammonium salt supplied as a high-purity solid for research use. It functions as an electrolyte and phase-transfer reagent and is used as a supporting salt in organic synthesis and analytical applications.

  • High purity: 98.0% assay.
  • Solid form: white powder or crystals.
  • Common applications include electrochemistry, phase-transfer catalysis, and analytical methods.
  • Molecular formula C4H12BF4N; molecular weight 160.95 g/mol.
  • CAS number 661-36-9 for unambiguous identification.
  • Packaged for laboratory use in 50 G quantities.

Catalog No. 50-002-82745 Supplier Medchemexpress LLC Supplier No. HYW12767250G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetramethylammonium tetrafluoroborate is an inorganic quaternary ammonium salt supplied as a high-purity solid for research use. It functions as an electrolyte and phase-transfer reagent and is used as a supporting salt in organic synthesis and analytical applications.

  • High purity: 98.0% assay.
  • Solid form: white powder or crystals.
  • Common applications include electrochemistry, phase-transfer catalysis, and analytical methods.
  • Molecular formula C4H12BF4N; molecular weight 160.95 g/mol.
  • CAS number 661-36-9 for unambiguous identification.
  • Packaged for laboratory use in 50 G quantities.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.