Learn More
Medchemexpress LLC Tetramethylammonium tetrafluoroborate | 661-36-9 | MFCD00011745 | 98.0% | 160.95 g/mol | C4H12BF4N | 500 G
SDP
Catalog No. 5000283521
Encompass_Preferred
Tetramethylammonium tetrafluoroborate is a white to off-white solid used as a biochemical reagent in laboratory research. It serves as a source of the tetramethylammonium cation paired with a non-coordinating tetrafluoroborate anion, and is suitable for use in assays, synthetic procedures, and as an electrolyte or phase-transfer agent where inert counterions are required.
  • High purity (≥98.0%) for reproducible results.
  • White to off-white solid, easy to weigh and handle.
  • Soluble in common organic solvents for versatile use.
  • Available in bulk pack sizes including 500 G for scale-up.
  • Stable when stored sealed and protected from moisture.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-83521 Supplier Medchemexpress LLC Supplier No. HYW127672500G
Tetramethylammonium tetrafluoroborate is a white to off-white solid used as a biochemical reagent in laboratory research. It serves as a source of the tetramethylammonium cation paired with a non-coordinating tetrafluoroborate anion, and is suitable for use in assays, synthetic procedures, and as an electrolyte or phase-transfer agent where inert counterions are required.
  • High purity (≥98.0%) for reproducible results.
  • White to off-white solid, easy to weigh and handle.
  • Soluble in common organic solvents for versatile use.
  • Available in bulk pack sizes including 500 G for scale-up.
  • Stable when stored sealed and protected from moisture.
  • Documentation available: data sheet, certificate of analysis, and safety data sheet.