Learn More
Crescent Chemical Co Inc TETRAMETHYLBENZIDINE (TMB)
SDP
Catalog No. 50248185
Encompass_Preferred
5,5'-Tetramethylbenzidine research grade Substrate for horseradish peroxidase. More sensitive than ABTS and more stable and less toxic than diaminobenzidine. See also TMB Ready-To-Use ELISA Substrate (cat. no. 37068) and SERVAColor TMB Blot Solution (cat. no. 37071). S
Catalog No. 50-248-185 Supplier Crescent Chemical Co Inc Supplier No. SE35926.02
5,5'-Tetramethylbenzidine research grade Substrate for horseradish peroxidase. More sensitive than ABTS and more stable and less toxic than diaminobenzidine. See also TMB Ready-To-Use ELISA Substrate (cat. no. 37068) and SERVAColor TMB Blot Solution (cat. no. 37071). S