Learn More
Selleck Chemical LLC Tetramethylcurcumin E2644-5MG
SDP
Catalog No. 5000040722
Encompass_Preferred
Tetramethylcurcumin (FLLL31) a small-molecule signal transducer and activator of transcription 3 (STAT3) inhibitor derived from curcumin binds selectively to Janus kinase 2 and the STAT3 Src homology-2 domain which serve crucial roles in STAT3 dimerization and signal transduction
Catalog No. 50-000-40722 Supplier Selleck Chemical LLC Supplier No. E26445MG
Tetramethylcurcumin (FLLL31) a small-molecule signal transducer and activator of transcription 3 (STAT3) inhibitor derived from curcumin binds selectively to Janus kinase 2 and the STAT3 Src homology-2 domain which serve crucial roles in STAT3 dimerization and signal transduction