Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetramethylcyclopentadiene, mixed isomers, tech. 85%

Catalog No. aa3857703
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3857703 1 g
AA3857706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3857703 Supplier Thermo Scientific Chemicals Supplier No. 03857703
Only null left
Add to Cart

CAS: 4249-10-9 | C9H14 | 122.21 g/mol

Tetramethylcyclopentadiene is used in pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4249-10-9
Molecular Formula C9H14
Molecular Weight (g/mol) 122.21
MDL Number MFCD00145385
InChI Key VNPQQEYMXYCAEZ-UHFFFAOYSA-N
Synonym 1,2,3,4-tetramethylcyclopentadiene, tetramethylcyclopentadiene, 1,2,3,4-tetramethyl-1,3-cyclopentadiene, tetramethyl cyclopentadiene, tetramethylcyclopentadiene, mixed isomers, pubchem15072, acmc-1an6q, 1,3-cyclopentadiene, tetramethyl, 2,3,4,5-tetramethylcyclopenta-2,4-diene
PubChem CID 138163
IUPAC Name 1,2,3,4-tetramethylcyclopenta-1,3-diene
SMILES CC1=C(C(=C(C1)C)C)C

Specifications

Color Colorless
Density 0.808 g/mL
Boiling Point 55°C to 59°C (13 mmHg)
Flash Point 42°C (108°F)
Refractive Index 1.472
Quantity 1 g
UN Number UN3295
Sensitivity Air and moisture sensitive
Solubility Information Not miscible mix with mater.
Formula Weight 122.21
Percent Purity 85%
Grade Technical
Physical Form Liquid
Chemical Name or Material Tetramethylcyclopentadiene, Mixed isomers
Show More Show Less
Hazard Category H226
Hazard Statement GHS H Statement
H226
Flammable liquid and vapor.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.