Learn More
Electron Microscopy Sciences (TMA), Tetramethylene Glycol Dimethacrylate 100 ML

Supplier: Electron Microscopy Sciences 21750
(TMA)
CAS #109-17-1 [H₂C=C₉CH₃)CO₂CH₂CH₂] ₂ F.W. 226.68
Boiling point: 132-134°C
(TMA), Tetramethylene Glycol Dimethacrylate (1,4-Butanediol dimethacrylate) inhibited with 200 ppm hydroquinone monomethyl ether.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.