Learn More
Medchemexpress LLC HY-N0264 100mg Medchemexpress, Tetramethylpyrazine CAS:1124-11-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0264100MG
Encompass_Preferred
Medchemexpress, HY-N0264 100mg Tetramethylpyrazine CAS:1124-11-4 Tetramethylpyrazine (Ligustrazine), an alkylpyrazine isolated from Ligusticum wallichii (Chuan Xiong) [1] , is present in french fries, bread, cooked meats, tea, cocoa, coffee, beer, spirits, peanuts, filberts, dairy products and soy products as fragrance and flavouring ingredienexhibits. Tetramethylpyrazine (Ligustrazine) also has potential nootropic and anti-inflammatory activities in rats [2] [3] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3499