Learn More
Cayman Chemical Tetramethylrhodamine methyl ester (perchlorate), Purity: ≥98%, CAS: 115532-50-8, 50 mg
SDP
Catalog No. 502994111
Encompass_Preferred
A cell-permeant fluorogenic dye used to assess mitochondrial function and cell viability; excitation/emission spectra 515 and 555/575 nm
Catalog No. 50-299-4111 Supplier Cayman Chemical Supplier No. 2143750 MG
A cell-permeant fluorogenic dye used to assess mitochondrial function and cell viability; excitation/emission spectra 515 and 555/575 nm