Learn More
Invitrogen™ Tetramethylrhodamine-5-Isothiocyanate (5-TRITC; G isomer)
Description
The amine-reactive tetramethylrhodamine-5-isothiocyanate (5-TRITC; G isomer) can be used to can be used to create bright red-orange fluorescent bioconjugates with excitation/emission maxima ∼555/580 nm.
Specifications
Specifications
| Quantity | 5 mg |
| Product Type | Tetramethylrhodamine-5-isothiocyanate |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Amine |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Isothiocyanate |
| Label or Dye | TMR (Tetramethylrhodamine), TRITC Isomers |
| Label Type | Classic Dyes |
| Emission | 580 |
| Excitation | 555 |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.