Learn More
Cayman Chemical Tetramethylrhodamine ethyl ester (perchlorate), Purity: ≥98%, CAS: 115532-52-0, 5 mg
SDP
Catalog No. 502994625
Encompass_Preferred
A cell-permeant fluorogenic dye used to assess mitochondrial function and cell viability; excitation/emission spectra 550/575 nm
Catalog No. 50-299-4625 Supplier Cayman Chemical Supplier No. 214265 MG
A cell-permeant fluorogenic dye used to assess mitochondrial function and cell viability; excitation/emission spectra 550/575 nm