Learn More
Medchemexpress LLC Tetramethylrhodamine methyl ester perchlorate | 115532-50-8 | MFCD00274460 | 99.7% | 500.93 g/mol | C25H25ClN2O7 | 10 MG
SDP
Catalog No. 5000297673
Encompass_Preferred
TMRM perchlorate is a red-fluorescent, membrane-permeant rhodamine dye used to measure mitochondrial membrane potential in live cells. It accumulates in active mitochondria in a membrane potential-dependent manner, enabling visualization and quantitative assessment of mitochondrial polarization for imaging and functional assays.
  • Red fluorescent probe with excitation at 550 nm and emission at 576 nm.
  • High purity (99.7%), suitable for research applications.
  • Membrane-permeant; accumulates in mitochondria in a membrane potential-dependent manner.
  • Suitable for live-cell imaging and quantitative mitochondrial membrane potential assays.
  • Provided as a 10 mg solid for laboratory use.
Catalog No. 50-002-97673 Supplier Medchemexpress LLC Supplier No. HYD0984A10MG
TMRM perchlorate is a red-fluorescent, membrane-permeant rhodamine dye used to measure mitochondrial membrane potential in live cells. It accumulates in active mitochondria in a membrane potential-dependent manner, enabling visualization and quantitative assessment of mitochondrial polarization for imaging and functional assays.
  • Red fluorescent probe with excitation at 550 nm and emission at 576 nm.
  • High purity (99.7%), suitable for research applications.
  • Membrane-permeant; accumulates in mitochondria in a membrane potential-dependent manner.
  • Suitable for live-cell imaging and quantitative mitochondrial membrane potential assays.
  • Provided as a 10 mg solid for laboratory use.