Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Tetramethylrhodamine methyl ester perchlorate

Catalog No. AC464560250
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC464560250 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AC464560250 Supplier Thermo Scientific Chemicals Supplier No. 464560250
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

CAS: 115532-50-8 | C25H25ClN2O7 | 500.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115532-50-8
Molecular Formula C25H25ClN2O7
Molecular Weight (g/mol) 500.93
InChI Key PFYWPQMAWCYNGW-UHFFFAOYSA-M
IUPAC Name 6-(dimethylamino)-9-[2-(methoxycarbonyl)phenyl]-N,N-dimethyl-3H-xanthen-3-iminium perchlorate
SMILES [O-][Cl](=O)(=O)=O.COC(=O)C1=CC=CC=C1C1=C2C=CC(C=C2OC2=CC(=CC=C12)N(C)C)=[N+](C)C

Specifications

Quantity 25 mg
Formula Weight 500.93
Chemical Name or Material Tetramethylrhodamine methyl ester perchlorate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.