Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetramethylthiuram monosulfide | 97-74-5 | MFCD00014870 | 99.7% | 208.37 | C6H12N2S3 | 50 MG
SDP

Catalog No. 5000299483
Encompass_Preferred

Tetramethylthiuram monosulfide is an organosulfur research reagent used in rubber research and various biochemical studies. It is a light yellow solid with molecular formula C6H12N2S3 and a molecular weight of 208.37 g/mol, supplied at high purity with defined storage recommendations for powder and solution forms.

  • High purity: 99.7%
  • Appearance: light yellow solid
  • Molecular formula: C6H12N2S3
  • Molecular weight: 208.37 g/mol
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years) or -20°C (1 year)
  • Applications: rubber research and various biochemical studies

Catalog No. 50-002-99483 Supplier Medchemexpress LLC Supplier No. HYW02024650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetramethylthiuram monosulfide is an organosulfur research reagent used in rubber research and various biochemical studies. It is a light yellow solid with molecular formula C6H12N2S3 and a molecular weight of 208.37 g/mol, supplied at high purity with defined storage recommendations for powder and solution forms.

  • High purity: 99.7%
  • Appearance: light yellow solid
  • Molecular formula: C6H12N2S3
  • Molecular weight: 208.37 g/mol
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (2 years) or -20°C (1 year)
  • Applications: rubber research and various biochemical studies

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.