Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetramethyltin

Catalog No. 48139450ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-139-450ML 50 mL
48-139-410ML 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 48-139-450ML Supplier Sigma Aldrich Supplier No. 48139450ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetramethyltin

Specifications

CAS 594-27-4
Melting Point -54°C
Density 1.291 g/mL (at 25°C)
Boiling Point 74°C to 75°C
Molecular Formula C4H12Sn
Refractive Index n20/D 1.441 (literature)
Linear Formula Sn(CH3)4
Quantity 50 mL
MDL Number MFCD00008278
Synonym Tin tetramethyl
Molecular Weight (g/mol) 178.85
Percent Purity 95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.