Learn More
Apexbio Technology LLC (-)-Tetramisole 16595-80-5 10g
SDP
Catalog No. 5000094680
Encompass_Preferred
(-)-Tetramisole is an immunomodulatory agent that stimulates nicotinic acetylcholine receptors leading to the activation of T-lymphocytes macrophages and phagocytic cells to enhance cell-mediated immune responses (-)-Tetramisole exerts its biological activity primarily through activation of nicotinic acetylcholine receptors Additionally it is utilized in the investigation of epigenetic regulation pathways involving lysine-specific demethylase 1 (LSD1) serving as a research tool to study chromatin modulation mechanisms and gene expression control Based on these pharmacological properties (-)-Tetramisole holds research potential in immunological signaling studies and tumor biology
Catalog No. 50-000-94680 Supplier Apexbio Technology LLC Supplier No. B177710000
(-)-Tetramisole is an immunomodulatory agent that stimulates nicotinic acetylcholine receptors leading to the activation of T-lymphocytes macrophages and phagocytic cells to enhance cell-mediated immune responses (-)-Tetramisole exerts its biological activity primarily through activation of nicotinic acetylcholine receptors Additionally it is utilized in the investigation of epigenetic regulation pathways involving lysine-specific demethylase 1 (LSD1) serving as a research tool to study chromatin modulation mechanisms and gene expression control Based on these pharmacological properties (-)-Tetramisole holds research potential in immunological signaling studies and tumor biology