Learn More
Apexbio Technology LLC (-)-Tetramisole 16595-80-5 5g
SDP
Catalog No. 502240558
Encompass_Preferred
(-)-Tetramisole is an immunomodulatory agent that stimulates nicotinic acetylcholine receptors leading to the activation of T-lymphocytes macrophages and phagocytic cells to enhance cell-mediated immune responses (-)-Tetramisole exerts its biological activity primarily through activation of nicotinic acetylcholine receptors Additionally it is utilized in the investigation of epigenetic regulation pathways involving lysine-specific demethylase 1 (LSD1) serving as a research tool to study chromatin modulation mechanisms and gene expression control Based on these pharmacological properties (-)-Tetramisole holds research potential in immunological signaling studies and tumor biology
This item is not returnable. View return policy
Catalog No. 50-224-0558 Supplier Apexbio Technology LLC Supplier No. B17775000
(-)-Tetramisole is an immunomodulatory agent that stimulates nicotinic acetylcholine receptors leading to the activation of T-lymphocytes macrophages and phagocytic cells to enhance cell-mediated immune responses (-)-Tetramisole exerts its biological activity primarily through activation of nicotinic acetylcholine receptors Additionally it is utilized in the investigation of epigenetic regulation pathways involving lysine-specific demethylase 1 (LSD1) serving as a research tool to study chromatin modulation mechanisms and gene expression control Based on these pharmacological properties (-)-Tetramisole holds research potential in immunological signaling studies and tumor biology