Learn More
Medchemexpress LLC HY-13764 100mg Medchemexpress, Tetrandrine CAS:518-34-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13764100MG
Encompass_Preferred
Medchemexpress, HY-13764 100mg Tetrandrine CAS:518-34-3 Tetrandrine (NSC-77037; d-Tetrandrine) is a bis-benzyl-isoquinoline alkaloid, which inhibits voltage-gated Ca2+ current (ICa) and Ca2+-activated K+ current. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9028