Learn More
Medchemexpress LLC Tetrandrine | 518-34-3 | 99.9% | 622.75 | 250 MG
SDP
Catalog No. 5000351112
Encompass_Preferred
Tetrandrine, also known as NSC-77037 or d-Tetrandrine, is a bis-benzyl-isoquinoline alkaloid. It functions as an inhibitor of voltage-gated Ca2+ current (ICa) and Ca2+-activated K+ current. This compound exhibits a broad spectrum of biological activities, including anti-inflammatory, anti-fibrogenic, anti-silicosis, anti-arthritis, and anti-cancer properties. It is also recognized as a potent calcium channel blocker, capable of regulating various cellular processes.
  • Inhibits voltage-gated Ca2+ and Ca2+-activated K+ currents
  • Possesses anti-inflammatory and anti-fibrogenic effects
  • Demonstrates anti-silicosis and anti-arthritis activities
  • Exhibits anti-cancer properties, including tumor growth inhibition and metastasis prevention
  • Can reverse multidrug resistance
  • Induces apoptosis and autophagy
  • Regulates cell proliferation, differentiation, and apoptosis
Catalog No. 50-003-51112 Supplier Medchemexpress LLC Supplier No. 250MGHY13764
Tetrandrine, also known as NSC-77037 or d-Tetrandrine, is a bis-benzyl-isoquinoline alkaloid. It functions as an inhibitor of voltage-gated Ca2+ current (ICa) and Ca2+-activated K+ current. This compound exhibits a broad spectrum of biological activities, including anti-inflammatory, anti-fibrogenic, anti-silicosis, anti-arthritis, and anti-cancer properties. It is also recognized as a potent calcium channel blocker, capable of regulating various cellular processes.
  • Inhibits voltage-gated Ca2+ and Ca2+-activated K+ currents
  • Possesses anti-inflammatory and anti-fibrogenic effects
  • Demonstrates anti-silicosis and anti-arthritis activities
  • Exhibits anti-cancer properties, including tumor growth inhibition and metastasis prevention
  • Can reverse multidrug resistance
  • Induces apoptosis and autophagy
  • Regulates cell proliferation, differentiation, and apoptosis