Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrandrine, ≥90%, MilliporeSigma™ Supelco™

Catalog No. 111014419 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-4419 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-4419 Supplier MilliporeSigma Supelco Supplier No. T26951G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetrandrine (TET) is classified under the bisbenzylisoquinoline alkaloid family of compounds extracted from Stephania tetrandra S. Moore.

TRUSTED_SUSTAINABILITY

Specifications

CAS 518-34-3
Physical Form Solid
Linear Formula C38H42N2O6
MDL Number MFCD08689909
Synonym (1 beta)-6,6′,7,12-Tetramethoxy-2,2′-dimethylberbaman
Type Analytical Standard
Industry Type Drug Analysis
Quantity 1 g
Formula Weight 622.75
Percent Purity ≥90%
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.