Learn More
Selleck Chemical LLC Tetrandrine 100mg 518-34-3 Fanchinine
SDP
Supplier:  Selleck Chemical LLC S2403100MG
Encompass_Preferred
Tetrandrine (Fanchinine), a bis-benzylisoquinoline alkaloid derived from Stephania tetrandra, is a calcium channel blocker. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-537-0