Learn More
Thermo Scientific Chemicals Tetranitro blue tetrazolium chloride, 97%
Description
Redox indicator for enzymes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Beilstein | 3897475 |
| Color | Yellow |
| Quantity | 100 mg |
| Physical Form | Powder |
| IUPAC Name | 2-{4'-[3,5-bis(4-nitrophenyl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium-2-yl]-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl}-3,5-bis(4-nitrophenyl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium dichloride |
| Formula Weight | 999.78 |
| Percent Purity | 97% |
| Chemical Name or Material | Tetranitro blue tetrazolium chloride |
Safety and Handling
| Hazard Category | H350 |
| Hazard Statement | GHS H Statement H350 May cause cancer. |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| EINECSNumber | 214-665-1 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.