Learn More
Cayman Chemical tetranorPGDMd6 GC MS Standards
SDP
Supplier:  Cayman Chemical 10009039100 UG
Encompass_Preferred
An internal standard for the quantification of tetranor-PGDM by GC- or LC-MS
Catalog No. 50-288-8969