Learn More
Cayman Chemical tetranorPGDMd6 GC MS Standards
SDP
Catalog No. 502888970
Encompass_Preferred
An internal standard for the quantification of tetranor-PGDM by GC- or LC-MS
Catalog No. 50-288-8970 Supplier Cayman Chemical Supplier No. 1000903925 UG
An internal standard for the quantification of tetranor-PGDM by GC- or LC-MS