Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetraoctylammonium Hydroxide Solution

Catalog No. 8799350ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
87-993-50ML 10 mL
87-993-10ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 87-993-50ML Supplier Sigma Aldrich Supplier No. 8799350ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetraoctylammonium hydroxide solution can be used as:A lipophilic base in the study of its effect on opticalpCO2sensors.A lipophilic organic base in the preparation of nanobeads, which are used as sensors for the detection of acidic and basic gases.A starting material for the preparation of ionic liquid named tetraoctylammonium 2-methyl-1-naphthoate, which is used in the extraction of 2,3-butanediol from H2O.Very lipophilic phase-transfer agent; employed in a carbon dioxide optical sensor

Specifications

CAS 17756-58-0
Molecular Formula C32H69NO
Linear Formula [CH3(CH2)7]4N(OH)
MDL Number MFCD00059257
Quantity 10 mL
Molecular Weight (g/mol) 483.9 g/mol
Biological Activity Eyes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.