Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetrapentylammonium iodide | 2498-20-6 | MFCD00041980 | >98.0% | 425.5 g/mol | C20H44IN | 10 G
SDP

Catalog No. 5000283606
Encompass_Preferred

Tetraamylammonium iodide is a quaternary ammonium iodide salt supplied for laboratory research use. It serves as a biochemical reagent and a source of tetraamylammonium cations in organic synthesis and ion-channel studies, provided as a solid for solution preparation and analytical applications.

  • High purity (>98.0%).
  • White to off-white crystalline solid.
  • Molecular formula C20H44IN.
  • Molecular weight 425.5 g/mol.
  • CAS number 2498-20-6.
  • Supplied in a 10 G package.
  • Intended for research use only.

Catalog No. 50-002-83606 Supplier Medchemexpress LLC Supplier No. HYW12765910G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetraamylammonium iodide is a quaternary ammonium iodide salt supplied for laboratory research use. It serves as a biochemical reagent and a source of tetraamylammonium cations in organic synthesis and ion-channel studies, provided as a solid for solution preparation and analytical applications.

  • High purity (>98.0%).
  • White to off-white crystalline solid.
  • Molecular formula C20H44IN.
  • Molecular weight 425.5 g/mol.
  • CAS number 2498-20-6.
  • Supplied in a 10 G package.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.