Learn More
Medchemexpress LLC Tetrapeptide-1 | 1632354-04-1 | 98.41% | 428.52 | 5 MG
SDP
Catalog No. 5000397995
Encompass_Preferred
Tetrapeptide-1 is a signaling peptide with antioxidant properties. It can stimulate the synthesis of extracellular matrices such as collagen, elastin, and fibronectin, thereby helping to combat signs of skin aging like wrinkles and sagging. It is suitable for use as a cosmetic ingredient.
  • Signaling peptide with antioxidant properties
  • Stimulates synthesis of extracellular matrices like collagen and elastin
  • Helps combat signs of skin aging
  • Suitable for use as a cosmetic ingredient
Catalog No. 50-003-97995 Supplier Medchemexpress LLC Supplier No. HYP52485MG
Tetrapeptide-1 is a signaling peptide with antioxidant properties. It can stimulate the synthesis of extracellular matrices such as collagen, elastin, and fibronectin, thereby helping to combat signs of skin aging like wrinkles and sagging. It is suitable for use as a cosmetic ingredient.
  • Signaling peptide with antioxidant properties
  • Stimulates synthesis of extracellular matrices like collagen and elastin
  • Helps combat signs of skin aging
  • Suitable for use as a cosmetic ingredient