Learn More
Strem, An Ascensus Company Ph4AsCl hydrate, 99% 5g
SDP
Supplier:  Strem, An Ascensus Company 3330505G
Encompass
CAS# 104170-16-3. 5g. Tetraphenylarsonium chloride monohydrate, 99%. MFCD00149995. Molecular Weight: 418.80 (436.81). Molecular Formula: (C6H5)4AsCl∙H2O. Color/form: white pwdr. Strem# 33-3050. http://www.strem.com/catalog/v/33-3050/
Catalog No. 50-901-11429