Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraphenylarsonium chloride hydrate, 96%, Thermo Scientific™

Catalog No. AAA1604303
Change view
Click to view available options
Quantity:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1604303 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1604303 Supplier Thermo Scientific Chemicals Supplier No. A1604303

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Tetraphenylarsonium chloride hydrate is a cation exchange reagent. It is used to prepare arsonium amide, oxime, and carbohydrate derivatives.

Solubility
Freely soluble in water. Soluble in alcohol. Sparingly soluble in acetone

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 507-28-8
Molecular Formula C24H20AsCl
Quantity 1g
MDL Number MFCD00011908
UN Number UN3465
Beilstein 3580063
Merck Index 14,9236
Synonym tetraphenylarsonium chloride, tetraphenylarsenium chloride, unii-ag51mkf20j, arsonium, tetraphenyl-, chloride, phenylarsonium chloride ph4ascl, ag51mkf20j, tetraphenylarsonium v chloride hydrate, phenylarsonium chloride, c24h20as.cl, ccris 9327
Solubility Information Freely soluble in water; Soluble in alcohol; Sparingly soluble in acetone
InChI Key NRZGVGVFPHPXEO-UHFFFAOYSA-M
SMILES C1=CC=C(C=C1)[As+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.[Cl-]
IUPAC Name tetraphenylarsanium;chloride
Molecular Weight (g/mol) 418.796
PubChem CID 68179
Formula Weight 418.80 (Anhydrous)
Percent Purity 96%
Chemical Name or Material Tetraphenylarsonium chloride hydrate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H301-H331
Toxic if swallowed.
Toxic if inhaled.
Precautionary Statement GHS P Statement
P261-P301+P310-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOARSENIC COMPOUND, SOLID, N.O.S.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.