Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraphenylarsonium Chloride Hydrochloride Hydrate 98.0+%, TCI America™
SDP

Catalog No. T01621G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T01621G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T01621G Supplier TCI America Supplier No. T01621G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 123334-18-9
Molecular Formula C24H23AsCl2O
Molecular Weight (g/mol) 473.269
MDL Number MFCD00012628
InChI Key HRCBNZHNCWEFFR-UHFFFAOYSA-M
PubChem CID 16211271
IUPAC Name tetraphenylarsanium;chloride;hydrate;hydrochloride
SMILES C1=CC=C(C=C1)[As+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.O.Cl.[Cl-]

Specifications

Color White-Yellow
Quantity 1 g
UN Number 3465
Formula Weight 455.25
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Tetraphenylarsonium Chloride Hydrochloride Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.