Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraphenylarsonium Chloride Monohydrate, For Analysis, EMSURE™, MilliporeSigma™

Catalog No. M1081500005 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M1081500005 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1081500005 Supplier MilliporeSigma Supplier No. 1.08150.0005

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 104170-16-3
Molecular Formula C24H22AsClO
Molecular Weight (g/mol) 436.811
InChI Key NGDWSDTZKCSLRK-UHFFFAOYSA-M
Synonym tetraphenylarsenic chloride, tetraphenylarsonium chloride monohydrate, arsonium, tetraphenyl-,chloride, hydrate 1:1:1, phenylarsonium chloride, tetraphenylarsanium chloride hydrate, acmc-20m6yd, c24h20as.cl.h2o, tetraphenyl-arsonium hydrate chloride, tetraphenylarsonium v chloride hydrate
PubChem CID 2723786
IUPAC Name tetraphenylarsanium;chloride;hydrate
SMILES C1=CC=C(C=C1)[As+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.O.[Cl-]

Specifications

Physical Form Solid
Quantity 5 g
UN Number UN 3465
Formula Weight 436.82. (molar mass)
Odor Odorless
Packaging Glass bottle
Appearance White solid
Chemical Name or Material Tetraphenylarsonium chloride monohydrate
Hazard Category H301 + H331: Toxic if swallowed or if inhaled
H410: Very toxic to aquatic life with long lasting effects.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.