Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraphenylarsonium Chloride Monohydrate p.a., Thermo Scientific™

Catalog No. AC138510025
Change view
Click to view available options
Quantity:
2.5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC138510025 2.5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC138510025 Supplier Thermo Scientific Chemicals Supplier No. 138510025

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104170-16-3
Molecular Formula C24H22AsClO
Molecular Weight (g/mol) 436.811
InChI Key NGDWSDTZKCSLRK-UHFFFAOYSA-M
Synonym tetraphenylarsenic chloride, tetraphenylarsonium chloride monohydrate, arsonium, tetraphenyl-,chloride, hydrate 1:1:1, phenylarsonium chloride, tetraphenylarsanium chloride hydrate, acmc-20m6yd, c24h20as.cl.h2o, tetraphenyl-arsonium hydrate chloride, tetraphenylarsonium v chloride hydrate
PubChem CID 2723786
IUPAC Name tetraphenylarsanium;chloride;hydrate
SMILES C1=CC=C(C=C1)[As+](C2=CC=CC=C2)(C3=CC=CC=C3)C4=CC=CC=C4.O.[Cl-]

Specifications

Color Beige
Infrared Spectrum Authentic
Linear Formula (C6H5)4AsCl.H2O
Quantity 2.5 g
Sensitivity (to tin and mercury) passes
Formula Weight 436.82
Percent Purity ≥99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tetraphenylarsonium chloride monohydrate, p.a.
Hazard Category GHS Signal Word: Danger
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.