Learn More
Medchemexpress LLC Tetraphenylethene maleimide | 1245606-71-6 | 98.9% | C31H23NO2 | 10MG
SDP
Catalog No. 5000261425
Encompass_Preferred
TPE-MI is a fluorogenic thiol probe used to measure unfolded protein load and proteostasis in cells. It exhibits aggregation-induced emission upon reaction with exposed thiols, providing a turn-on fluorescence signal suitable for cellular studies of protein folding and proteostasis-related disease models.
  • Fluorogenic thiol probe that reports exposed cysteines.
  • Aggregation-induced emission gives a strong turn-on signal.
  • Excitation 350 nm, emission 470 nm for fluorescence detection.
  • High purity (98.93%) solid with light yellow to yellow appearance.
  • Available in multiple small package sizes for research use.
Catalog No. 50-002-61425 Supplier Medchemexpress LLC Supplier No. HY14321810MG
TPE-MI is a fluorogenic thiol probe used to measure unfolded protein load and proteostasis in cells. It exhibits aggregation-induced emission upon reaction with exposed thiols, providing a turn-on fluorescence signal suitable for cellular studies of protein folding and proteostasis-related disease models.
  • Fluorogenic thiol probe that reports exposed cysteines.
  • Aggregation-induced emission gives a strong turn-on signal.
  • Excitation 350 nm, emission 470 nm for fluorescence detection.
  • High purity (98.93%) solid with light yellow to yellow appearance.
  • Available in multiple small package sizes for research use.