Learn More
Medchemexpress LLC Tetraphenylporphyrin | 917-23-7 | MFCD00011680 | 99.4% | 614.74 g/mol | C44H30N4 | 1 G
SDP
Catalog No. 5000246835
Encompass_Preferred
Tetraphenylporphyrin is a symmetrically substituted porphyrin-based heterocyclic compound provided as a solid research reagent for synthetic, photophysical, and materials studies.
  • High purity solid (99.4%) suitable for analytical and synthetic applications.
  • Common building block for supramolecular synthesis and materials research.
  • Well-characterized molecular formula C44H30N4 and molecular weight 614.74 g/mol.
  • Available in small pack sizes for laboratory-scale work.
  • Stable solid form, convenient for storage and handling.
Catalog No. 50-002-46835 Supplier Medchemexpress LLC Supplier No. HY1036561G
Tetraphenylporphyrin is a symmetrically substituted porphyrin-based heterocyclic compound provided as a solid research reagent for synthetic, photophysical, and materials studies.
  • High purity solid (99.4%) suitable for analytical and synthetic applications.
  • Common building block for supramolecular synthesis and materials research.
  • Well-characterized molecular formula C44H30N4 and molecular weight 614.74 g/mol.
  • Available in small pack sizes for laboratory-scale work.
  • Stable solid form, convenient for storage and handling.