Learn More
Medchemexpress LLC Tetraphenylporphyrin | 917-23-7 | MFCD00011680 | 99.4% | 614.74 g/mol | C44H30N4 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY103656500MG
Encompass_Preferred
Tetraphenylporphyrin (TPP) is a symmetrically substituted porphyrin used as a research reagent and building block in supramolecular synthesis, materials chemistry, and photophysical studies. It is supplied as a solid with high reported purity and defined molecular properties, suitable for laboratory-scale experiments.
  • High purity for research applications.
  • Solid, purple to black, easy to handle.
  • Widely used as a porphyrin building block in synthesis and supramolecular assemblies.
  • Compatible with photophysical and catalytic studies.
  • Stable when stored according to recommendations; protect from light.
Catalog No. 50-002-46836