Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetraphenyltin 97%, Thermo Scientific™

Catalog No. AC138591000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC138591000 100 g
AC138595000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC138591000 Supplier Thermo Scientific Chemicals Supplier No. 138591000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 595-90-4
Molecular Formula C24H20Sn
Molecular Weight (g/mol) 427.13
MDL Number MFCD00003006
InChI Key CRHIAMBJMSSNNM-UHFFFAOYSA-N
Synonym tetraphenyltin, stannane, tetraphenyl, tetraphenyl tin, ccris 6326, tin, tetraphenyl-,, stannane,tetraphenyl, acmc-209mem
PubChem CID 61146
IUPAC Name tetraphenylstannane
SMILES C1=CC=C(C=C1)[Sn](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 224°C to 228°C
Color White
Infrared Spectrum Authentic
Linear Formula Sn(C6H5)4
Quantity 100 g
Formula Weight 427.1
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material Tetraphenyltin, 97%
Hazard Category GHS Signal Word: Danger
EINECSNumber 209-872-9
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.