Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrapropylammonium bromide, 98%

Catalog No. AC209370025
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC209370025 2.5 kg Plastic bottle
AC209370250 25 g Plastic bottle
AC209371000 100 g Plastic bottle
AC209375000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC209370025 Supplier Thermo Scientific Chemicals Supplier No. 209370025
Only null left
Add to Cart

CAS: 1941-30-6 | C12H28BrN | 266.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1941-30-6
Molecular Formula C12H28BrN
Molecular Weight (g/mol) 266.26
MDL Number MFCD00011840
InChI Key BGQMOFGZRJUORO-UHFFFAOYSA-M
Synonym tetrapropylammonium bromide, tetra-n-propylammonium bromide, 1-propanaminium, n,n,n-tripropyl-, bromide, tetrapropylazanium bromide, tpabr, n,n,n-tripropyl-1-propanaminium bromide, n,n,n-tripropylpropan-1-aminium bromide, tetrapropylammonium bromide tpabr, 1-propanaminium, n,n,n-tripropyl-, bromide 1:1, tetra propyl ammonium bromide
PubChem CID 74745
ChEBI CHEBI:55318
IUPAC Name tetrapropylazanium;bromide
SMILES CCC[N+](CCC)(CCC)CCC.[Br-]

Specifications

Melting Point 266°C to 272°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5 to 102.5% (Argentometry)
Packaging Plastic bottle
Linear Formula (CH3CH2CH2)4NBr
Quantity 2.5 kg
Beilstein 04,I,364
Solubility Information Solubility in water: 100g/L water
Formula Weight 266.26
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material Tetrapropylammonium bromide, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 217-727-6
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.