Learn More
Medchemexpress LLC Tetrapropylammonium chloride | 5810-42-4 | ≥98.0% | 221.81 | 5 G
SDP
Catalog No. 5000452965
Encompass_Preferred
Tetrapropylammonium chloride is a quaternary ammonium compound with cationic properties. It serves as a biochemical reagent for life science research and finds application in various industrial and biomedical fields.
  • Used as a phase transfer catalyst in organic synthesis reactions.
  • Acts as a surfactant in personal care products, detergents, textiles, and pharmaceuticals.
  • Applied in the production of polymers and resins.
Catalog No. 50-004-52965 Supplier Medchemexpress LLC Supplier No. HYW0124125G
Tetrapropylammonium chloride is a quaternary ammonium compound with cationic properties. It serves as a biochemical reagent for life science research and finds application in various industrial and biomedical fields.
  • Used as a phase transfer catalyst in organic synthesis reactions.
  • Acts as a surfactant in personal care products, detergents, textiles, and pharmaceuticals.
  • Applied in the production of polymers and resins.