Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrapropylammonium Hydroxide Solution

Catalog No. 254533100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
25-453-3100G 100 g
25-453-325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Danger
Catalog No. 25-453-3100G Supplier Sigma Aldrich Supplier No. 254533100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetrapropylammonium hydroxide, a quaternary ammonium compound, is mainly used as a structure directing agent for the preparation of molecular sieves and zeolites.

Specifications

CAS 4499-86-9
Density 1.012 g/mL at 25°C
Molecular Formula C12H29NO
Linear Formula (CH3CH2CH2)4N(OH)
Refractive Index n20/D 1.3716
MDL Number MFCD00009360
Quantity 100 g
Synonym TPAOH solution
Molecular Weight (g/mol) 203.36 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.