Learn More
Medchemexpress LLC Tetrapropylammonium iodide | 631-40-3 | 313.26 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW009202
Encompass_Preferred
Tetrapropylammonium iodide is an organic compound widely utilized as an ionic liquid and catalyst. It serves multiple roles, including acting as a solvent in various chemical reactions and promoting organic synthesis reactions. Beyond its use in chemical synthesis, this compound finds extensive application in batteries, solar cells, and pigments.
  • Organic compound
  • Ionic liquid
  • Catalyst
  • Solvent
  • Organic synthesis
  • Broad applications
  • Biochemical reagent
Catalog No. 50-004-51778