Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tetrapropylammonium iodide, 98+%, Thermo Scientific™

Catalog No. AC216691000
Change view
Click to view available options
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC216691000 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC216691000 Supplier Thermo Scientific Chemicals Supplier No. 216691000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Specifications

CAS 631-40-3
Infrared Spectrum Authentic
Assay Percent Range 98+%
Molecular Formula C12H28IN
Linear Formula (CH3CH2CH2)4NI
Quantity 100g
MDL Number MFCD00011842
Beilstein 04, 140
Synonym tetrapropylammonium iodide, tetra-n-propylammonium iodide, ammonium, tetrapropyl-, iodide, tetrapropyl ammonium iodide, tetrapropylazanium iodide, n,n,n-tripropyl-1-propanaminium iodide, 1-propanaminium, n,n,n-tripropyl-, iodide, 1-propanaminium, n,n,n-tripropyl-, iodide 1:1, tetrapropylammoniumiodide, pr4ni
InChI Key GKXDJYKZFZVASJ-UHFFFAOYSA-M
SMILES CCC[N+](CCC)(CCC)CCC.[I-]
IUPAC Name tetrapropylazanium;iodide
Molecular Weight (g/mol) 313.267
PubChem CID 12429
Formula Weight 313.26
Percent Purity ≥98.0%
Chemical Name or Material Tetrapropylammonium iodide, 98+%
Show More Show Less
Hazard Category HYGROSCOPIC
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 211-157-1
RTECSNumber BS8410000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.