Learn More
Strem, An Ascensus Company CAS# 114615-82-6. 2g. Tetrapropylammonium perruthenate, min. 97% TPAP. MFCD00074914
SDP
Catalog No. 5090112030
Encompass
CAS# 114615-82-6. 2g. Tetrapropylammonium perruthenate, min. 97% TPAP. MFCD00074914. Molecular Weight: 351.43. Molecular Formula: (C3H7)4N+RuO4ae. Color/form: green xtl. Strem# 44-7850. http://www.strem.com/catalog/v/44-7850/
Catalog No. 50-901-12030 Supplier Strem, An Ascensus Company Supplier No. 4478502G
CAS# 114615-82-6. 2g. Tetrapropylammonium perruthenate, min. 97% TPAP. MFCD00074914. Molecular Weight: 351.43. Molecular Formula: (C3H7)4N+RuO4ae. Color/form: green xtl. Strem# 44-7850. http://www.strem.com/catalog/v/44-7850/