Learn More
Medchemexpress LLC Tetrapropylammonium perruthenate | 114615-82-6 | 351.43 | 1 G
SDP
Catalog No. 5000448595
Encompass_Preferred
Tetrapropylammonium perruthenate (TPAP) is an organic compound that serves as a catalyst and oxidizing agent. It is utilized in various organic synthesis reactions, particularly in catalyzing the oxidation of olefins and aromatic compounds. Beyond synthesis, TPAP finds broad application in industrial processes such as plastics, rubber, and textile manufacturing. It also functions as a biochemical reagent for life science research.
  • Functions as a catalyst
  • Acts as an oxidizing agent
  • Catalyzes oxidation of olefins and aromatic compounds
  • Used in plastics, rubber, and textile manufacturing
  • Applicable as a biochemical reagent for life science research
Catalog No. 50-004-48595 Supplier Medchemexpress LLC Supplier No. HYW0078011G
Tetrapropylammonium perruthenate (TPAP) is an organic compound that serves as a catalyst and oxidizing agent. It is utilized in various organic synthesis reactions, particularly in catalyzing the oxidation of olefins and aromatic compounds. Beyond synthesis, TPAP finds broad application in industrial processes such as plastics, rubber, and textile manufacturing. It also functions as a biochemical reagent for life science research.
  • Functions as a catalyst
  • Acts as an oxidizing agent
  • Catalyzes oxidation of olefins and aromatic compounds
  • Used in plastics, rubber, and textile manufacturing
  • Applicable as a biochemical reagent for life science research