Learn More
MOLTOX Molecular Toxicology Inc Tetrathionate Broth, 1000 mL/bottle, Store in Refrigerator
SDP
Catalog No. 50948849
Encompass_Preferred
Tetrathionate Broth is a selective medium used for the isolation of Salmonella from various materials such as food, urine, feces, etc. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-849 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 5140S120
Tetrathionate Broth is a selective medium used for the isolation of Salmonella from various materials such as food, urine, feces, etc. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.