Learn More
Alkali Scientific Tetrathionate Broth Base 10mL Tube, 10/Pk
SDP
Catalog No. 5000662457
Encompass_Preferred
Tetrathionate Broth Base is a preparatory medium used for the selective enrichment of Salmonella species. It contains essential nutrients and components required to generate tetrathionate after activation with an iodine solution. This activation step converts thiosulfate into tetrathionate, which selectively supports Salmonella growth while inhibiting competing bacteria. The base formulation allows laboratories to prepare fresh active medium immediately before use, ensuring optimal performance and reliability. This medium is commonly used in microbiology laboratories for food safety testing and environmental monitoring where accurate detection of Salmonella is required.
Catalog No. 50-006-62457 Supplier Alkali Scientific Supplier No. TTET01
Tetrathionate Broth Base is a preparatory medium used for the selective enrichment of Salmonella species. It contains essential nutrients and components required to generate tetrathionate after activation with an iodine solution. This activation step converts thiosulfate into tetrathionate, which selectively supports Salmonella growth while inhibiting competing bacteria. The base formulation allows laboratories to prepare fresh active medium immediately before use, ensuring optimal performance and reliability. This medium is commonly used in microbiology laboratories for food safety testing and environmental monitoring where accurate detection of Salmonella is required.