Learn More
HIMEDIA Tetrathionate HiVeg Broth Base, Hajna, MV327, Microbiology Culture Media, Dehydrated Culture Media, Selective Broth Base, Animal-Component-Free, Salmonella Enrichment, Powder, 500 g
Catalog No. NC4013779
Encompass_Preferred
Tetrathionate HiVeg Broth Base, Hajna is an animal-component-free selective broth for the enrichment of Salmonella from food and clinical specimens. Built on the plant-based HiVeg system, it uses vegetable peptones in place of animal peptones. An iodine-iodide solution added before use generates tetrathionate, which Salmonella tolerate and use while competing coliforms and other flora are held back. The Hajna formulation improves recovery and downstream selective isolation. Dehydrated powder, store in a cool dry place. Packaged as 500 g.
Catalog No. NC4013779 Supplier HIMEDIA Supplier No. MV327500G
Tetrathionate HiVeg Broth Base, Hajna is an animal-component-free selective broth for the enrichment of Salmonella from food and clinical specimens. Built on the plant-based HiVeg system, it uses vegetable peptones in place of animal peptones. An iodine-iodide solution added before use generates tetrathionate, which Salmonella tolerate and use while competing coliforms and other flora are held back. The Hajna formulation improves recovery and downstream selective isolation. Dehydrated powder, store in a cool dry place. Packaged as 500 g.