Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetrazine-PEG4-amine hydrochloride | 2752945-36-9 | 99.8% | 470.95 g/mol | C20H31ClN6O5 | 5 MG
SDP

Catalog No. 5000203366
Encompass_Preferred

Tetrazine-PEG4-amine hydrochloride is a cleavable four-unit polyethylene glycol (PEG) linker containing a tetrazine functional group used as a click-chemistry reagent for inverse electron-demand Diels-Alder (iEDDA) reactions and in the synthesis of antibody-drug conjugates. For research use only.

  • Cleavable four-unit PEG linker for ADC synthesis.
  • Tetrazine group enables fast iEDDA click chemistry with TCO partners.
  • Solid form with high purity (≈99.8%).
  • Molecular weight 470.95 g/mol; formula C20H31ClN6O5.
  • Soluble in methanol (≈250 mg/mL); may require sonication.
  • Store sealed at -20°C; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).

Catalog No. 50-002-03366 Supplier Medchemexpress LLC Supplier No. HY1309775MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetrazine-PEG4-amine hydrochloride is a cleavable four-unit polyethylene glycol (PEG) linker containing a tetrazine functional group used as a click-chemistry reagent for inverse electron-demand Diels-Alder (iEDDA) reactions and in the synthesis of antibody-drug conjugates. For research use only.

  • Cleavable four-unit PEG linker for ADC synthesis.
  • Tetrazine group enables fast iEDDA click chemistry with TCO partners.
  • Solid form with high purity (≈99.8%).
  • Molecular weight 470.95 g/mol; formula C20H31ClN6O5.
  • Soluble in methanol (≈250 mg/mL); may require sonication.
  • Store sealed at -20°C; in solution store at -80°C (up to 6 months) or -20°C (up to 1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.