Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Tetrazine-Peg5-Nhs Ester

Catalog No. 90091350MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-091-350MG 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 90-091-350MG Supplier Sigma Aldrich Supplier No. 90091350MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Tetrazine-PEG5-NHS ester has an N-hydroxysuccinimide (NHS) ester that can be reacted with primary amines and tetrazine, which is reactive to trans-cyclooctenes. Hydrogen substituted tetrazines demonstrate exceptionally fast kinetics, generally at least 10-fold faster compared to methyl substituted tetrazines. The aqueous solubility of this reagent is substantially enhanced by a hydrophilic polyethylene glycol (PEG) spacer arm.Tetrazine-PEG5-NHS ester (Tz-PEG5-NHS) can be used in click chemistry applications such as the preparation of chitosan-polyacrylamide microspheres utilizing selective tetrazine-trans-cyclooctene (Tz-TCO) ligation and the construction of copper-64 radiolabeled antibody by the click reaction.

Specifications

CAS 24367-66-6
Molecular Formula C33H51P
Linear Formula C33H51P
Quantity 1 g
Synonym Cyclopentylbis(3,5-di-tert-butylphenyl)phosphine; Phosphine ligand for nickel catalysis
Molecular Weight (g/mol) 478.73 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.