Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Tetrazine-ph-acid | 1380500-92-4 | MFCD24503098 | 98.7% | 216.2 | C10H8N4O2 | 100MG
SDP

Catalog No. 5000239338
Encompass_Preferred

Tetrazine-Ph-acid is a tetrazine-containing small molecule used as a click-chemistry reagent and PROTAC linker. It reacts rapidly via inverse electron-demand Diels-Alder (iEDDA) with trans-cyclooctene (TCO) derivatives, enabling fast bioorthogonal conjugation for synthesis and labeling applications.

  • Used as a PROTAC linker and click-chemistry reagent.
  • Undergoes inverse electron-demand Diels-Alder reactions with TCO derivatives.
  • Molecular formula C10H8N4O2; molecular weight 216.2; CAS 1380500-92-4.
  • Purity 98.7% (manufacturer-reported).
  • Appearance powder; recommended storage: powder at -20°C for long-term stability.
  • Soluble in DMSO (50 mg/mL); formulation solubility ≥2.5 mg/mL in recommended vehicles.
  • Supplied for research use; not intended for diagnostic or therapeutic use.

Catalog No. 50-002-39338 Supplier Medchemexpress LLC Supplier No. HY124480100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Tetrazine-Ph-acid is a tetrazine-containing small molecule used as a click-chemistry reagent and PROTAC linker. It reacts rapidly via inverse electron-demand Diels-Alder (iEDDA) with trans-cyclooctene (TCO) derivatives, enabling fast bioorthogonal conjugation for synthesis and labeling applications.

  • Used as a PROTAC linker and click-chemistry reagent.
  • Undergoes inverse electron-demand Diels-Alder reactions with TCO derivatives.
  • Molecular formula C10H8N4O2; molecular weight 216.2; CAS 1380500-92-4.
  • Purity 98.7% (manufacturer-reported).
  • Appearance powder; recommended storage: powder at -20°C for long-term stability.
  • Soluble in DMSO (50 mg/mL); formulation solubility ≥2.5 mg/mL in recommended vehicles.
  • Supplied for research use; not intended for diagnostic or therapeutic use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.